(3-Mercaptopropyl)trimethoxysilane ≥96.0%
Supplier: TCI America
Synonyms:
3-(Trimethoxysilyl)propane-1-thiol
CAS Number: 4420-74-0
MDL Number: MFCD00004901
Molecular Formula: C6H16O3SSi
Molecular Weight: 196.34
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 112
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Trimethoxysilyl)propanethiol
Formula:
HSH₂CCH₂CH₂Si(OCH₃)₃ MW: 196.34 g/mol Boiling Pt: 215 °C Melting Pt: –50 °C Density: 1.06 (20 °C) Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004901 CAS Number: 4420-74-0 EINECS: 224-588-5 UN: 3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...