2-Mercapto-1-methylimidazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Mercapto-1-methyl-1H-imidazole
CAS Number: 60-56-0
MDL Number: MFCD00179321
Molecular Formula: C4H6N2S
Molecular Weight: 114.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 145
Flash Point (°C): 110
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Methimazole, 1-Methylimidazole-2-thiol, Thiamazole
Formula:
C₄H₆N₂S MW: 114.17 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: 145 °C Density: 1.36 g/cm³ (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00179321 CAS Number: 60-56-0 EINECS: 200-482-4 Merck Index: 13,06000 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...