(±)-3-Methyl-1-pentanol ≥99.0%

Supplier: TCI America

Synonyms: (±)-3-Methylpentan-1-ol3-Methyl-1-pentanol3-Methylpentan-1-ol

M0600-100ML M0600-10ML
TCM0600-100MLEA 152.14 USD
TCM0600-100ML TCM0600-010ML
(±)-3-Methyl-1-pentanol ≥99.0%
(±)-3-Methyl-1-pentanol

CAS Number: 589-35-5
MDL Number: MFCD00002937
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 58
Specific Gravity (20/20): 0.83

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 152 °C
Melting Pt: –70 °C
Density: 0.83 (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002937
CAS Number: 589-35-5
EINECS: 209-644-9
UN: 1987
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/40/60/28974060.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR