(±)-Mandelonitrile ≥97.0%

Supplier: TCI America

M0566-25G
TCM0566-25GEA 30.62 USD
TCM0566-25G
(±)-Mandelonitrile ≥97.0%
(±)-Mandelonitrile

CAS Number: 532-28-5
MDL Number: MFCD00004487
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 97
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: 34.4 deg (C=5.3, Benzene)


Other Product Names / Synonyms: Benzaldehyde Cyanohydrin, alpha-Hydroxyphenylacetonitrile

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 102 °C
Density: 1.12 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00004487
CAS Number: 532-28-5
EINECS: 208-532-7
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/40/25/28974025.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR