1-Methylimidazole (NMI) ≥99.0%

Supplier: TCI America
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

M0508-500G M0508-25G M0508-100G
TCM0508-500GEA 125.46 USD
TCM0508-500G TCM0508-25G TCM0508-100G
1-Methylimidazole (NMI) ≥99.0%
1-Methylimidazole

CAS Number: 616-47-7 MDL Number: MFCD00005292 Molecular Formula: C4H6N2 Molecular Weight: 82.11 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 198 Flash Point (°C): 92 Specific Gravity (20/20): 1.04

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198 °C
Melting Pt: –2 °C
Density: 1.04 (20 °C)
Flash Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 3267
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14821e24b4b2a2910de1cdc82c9df9f974cbc8d&vid=b14821e24b4b2a2910de1cdc82c9df9f974cbc8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR