Morpholine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
M0465-500ML
M0465-25ML
TCM0465-500MLEA
50.1
USD
TCM0465-500ML
TCM0465-25ML
Morpholine ≥99.0% (by GC, titration analysis)
Morpholine
CAS Number: 110-91-8
MDL Number: MFCD00005972
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): -5
Flash Point (°C): 32
Specific Gravity (20/20): 1.00
Formula:
C₄H₉NO MW: 87.12 g/mol Boiling Pt: 129 °C Melting Pt: –5 °C Density: 1.00 (20 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005972 CAS Number: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8,I Merck Index: 12,06362 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...