3-Methylthiophene ≥98.0%

Supplier: TCI America

M0440-500G M0440-25G
TCM0440-500GEA 390 USD
TCM0440-500G TCM0440-25G
3-Methylthiophene ≥98.0%
3-Methylthiophene

CAS Number: 616-44-4 MDL Number: MFCD00005470 Molecular Formula: C5H6S Molecular Weight: 98.16 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 115 Flash Point (°C): 11 Specific Gravity (20/20): 1.02

Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 114
Melting Pt: -69
Density: 1.02 (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00005470
CAS Number: 616-44-4
EINECS: 210-482-6
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cb8bb942454346771415cda596d266c5452b67&vid=63cb8bb942454346771415cda596d266c5452b67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR