1-Methylpyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Methylpyrrolidine

M0415-500ML M0415-25ML M0415-100ML
TCM0415-100MLEA 71.8 USD
TCM0415-100ML TCM0415-25ML TCM0415-500ML
1-Methylpyrrolidine ≥98.0% (by GC, titration analysis)
1-Methylpyrrolidine

CAS Number: 120-94-5
MDL Number: MFCD00003173
Molecular Formula: C5H11N
Molecular Weight: 85.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 3
Specific Gravity (20/20): 0.80

Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 76 °C
Melting Pt: –76 °C
Density: 0.80 (20 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00003173
CAS Number: 120-94-5
EINECS: 204-438-5
UN: 3286
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/38/42/28973842.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR