1-Methylnaphthalene ≥96.0%

Supplier: TCI America
Warning

M0371-500ML M0371-25ML M0371-100ML
TCM0371-500MLEA 79.5 USD
TCM0371-500ML TCM0371-25ML TCM0371-100ML
1-Methylnaphthalene ≥96.0%
1-Methylnaphthalene

CAS Number: 90-12-0 MDL Number: MFCD00004034 Molecular Formula: C11H10 Molecular Weight: 142.20 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 241 Flash Point (°C): 82 Specific Gravity (20/20): 1.03

Formula: C₁₁H₁₀
Boiling Pt: 241 °C
Melting Pt: –33…–32 °C
Density: 1.03 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd72a505824185e056e3d78d9f94c41eb4a497a0&vid=dd72a505824185e056e3d78d9f94c41eb4a497a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR