Methanesulfonic acid ≥99.0%
Supplier: TCI America
|
Danger
|
M0093-500G
M0093-25G
TCM0093-500GEA
101.36
USD
TCM0093-500G
TCM0093-25G
Methanesulfonic acid ≥99.0%
Methanesulphonic acid
CAS Number: 75-75-2
MDL Number: MFCD00007518
Molecular Formula: CH4O3S
Molecular Weight: 96.10
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: Methylsulfonic Acid, Sulfomethane, MSA
Formula:
CH₃SO₃H Boiling Pt: 122 °C Melting Pt: 20 °C Density: 1.48 (20 °C) Flash Pt: 233 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007518 CAS Number: 75-75-2 EINECS: 200-898-6 UN: 2586 ADR: 8,III Merck Index: 13,05981 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...