Butyl methacrylate ≥99.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

M0081-500ML M0081-25ML
TCM0081-500MLEA 31.23 USD
TCM0081-500ML TCM0081-25ML
Butyl methacrylate ≥99.0% stabilized
Butyl methacrylate

(stabilized with HQ) CAS Number: 97-88-1 MDL Number: MFCD00009444 Molecular Formula: C8H14O2 Molecular Weight: 142.20 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 164 Melting point (°C): -25 Flash Point (°C): 51 Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS) Other Product Names / Synonyms: Methacrylic Acid Butyl Ester

Formula: C₈H₁₄O₂
Boiling Pt: 164 °C
Melting Pt: –25 °C
Density: 0.90 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00009444
CAS Number: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2305e62130b6fcb7333e0b1315debb3dd53ee8c&vid=d2305e62130b6fcb7333e0b1315debb3dd53ee8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR