2-Mercapto-2-thiazoline ≥98.0%
Supplier: TCI America
Synonyms:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
M0065-500G
M0065-25G
TCM0065-500GEA
127
USD
TCM0065-500G
TCM0065-25G
2-Mercapto-2-thiazoline ≥98.0%
2-Mercapto-2-thiazoline
CAS Number: 96-53-7
MDL Number: MFCD00126013
Molecular Formula: C3H5NS2
Molecular Weight: 119.20
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Thiazoline-2-thiol
Formula:
C₃H₅NS₂ Melting Pt: 105 °C |
MDL Number:
MFCD00126013 CAS Number: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...