Methyl orange 0.1% in water

Supplier: TCI America

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

M0017-500ML
TCM0017-500MLEA 56.95 USD
TCM0017-500ML
Methyl orange 0.1% in water
Methyl orange

[for Titration]
CAS Number: 547-58-0
MDL Number: MFCD00007502
Molecular Weight: 327.33
Form: Clear Liquid
Color: Red

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/32/78/28973278.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR