Light green SF yellowish

Supplier: TCI America

Synonyms: CI 42095Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonateDihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium saltSodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonateacid Green FLight Green 2G, S or 2GNDihydrogen (ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulfonatobenzyl)ammonium disodium saltacid Green 5Fast acid Green NCN DcL-30Food Green n°2

L0141-1G
TCL0141-001GEA 25.05 USD
TCL0141-001G
Light green SF yellowish
Light Green SF Yellowish

[as an indicator for assay of Food Yellow No.4 (Tartrazine)]
CAS Number: 5141-20-8
MDL Number: MFCD00012121
Molecular Formula: C37H36N2O9S3
Molecular Weight: 792.84
Form: Crystal
Color: Red

Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
Melting Pt: 288…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00012121
CAS Number: 5141-20-8
EINECS: 225-906-5
Merck Index: 13,05506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR