D(-)-Lysine monohydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
(R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride
CAS Number: 7274-88-6
MDL Number: MFCD00012920
Molecular Formula: C6H14N2O2
Molecular Weight: 182.65
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Lys-OH.HCl
Formula:
C₆H₁₅ClN₂O₂ Melting Pt: 261…262 °C |
MDL Number:
MFCD00012920 CAS Number: 7274-88-6 EINECS: 230-691-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...