Isobutanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: 2-Methyl-1-propanolIsobutylalcoholiso-Butanoliso-Butyl alcohol

I0094-25ML I0094-500ML
TCI0094-500MLEA 44.56 USD
TCI0094-500ML TCI0094-25ML
Isobutanol ≥99.0%
Isobutanol

CAS Number: 78-83-1
MDL Number: MFCD00004740
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Melting point (°C): -108
Flash Point (°C): 30
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: Isobutanol, Isobutyl Alcohol

Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 108 °C
Melting Pt: –108 °C
Density: 0.80 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://us.vwr.com/stibo/web/std.lang.all/18/80/28971880.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR