(±)-2-Aminoheptane ≥97.0%

Supplier: TCI America
Danger

Synonyms: 2-Aminoheptane, (±)-Tuaminoheptane, 1-Methylhexylamine, 2-Heptylamine, (±)-1-Methylhexylamine, Tuaminoheptane

H0645-25ML H0645-250ML
TCH0645-025MLEA 79.38 USD
TCH0645-025ML TCH0645-250ML
(±)-2-Aminoheptane ≥97.0%
(±)-2-Aminoheptane

CAS Number: 123-82-0 MDL Number: MFCD00008101 Molecular Formula: C7H17N Molecular Weight: 115.22 Purity/Analysis Method: 97.0% (GC,T) Form: Clear Liquid Boiling point (°C): 142 Flash Point (°C): 54 Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: 2-Aminoheptane

Formula: C₇H₁₇N
MW: 115.22 g/mol
Boiling Pt: 143
Density: 0.77 (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008101
CAS Number: 123-82-0
EINECS: 204-655-5
UN: 2734
ADR: 8 II
Merck Index: 12,09934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61715ffa313ba9856577697fc794abd0ce3771ab&vid=61715ffa313ba9856577697fc794abd0ce3771ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR