2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥95.0%

Supplier: TCI America
Danger

Synonyms: 2,2,3,3,4,4,4-Heptafluorobutan-1-ol 1H,1H-Perfluoro-1-butanol

H0548-25G H0548-5G
TCH0548-005GEA 57.98 USD
TCH0548-005G TCH0548-025G
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥95.0%
2,2,3,3,4,4,4-Heptafluoro-1-butanol

CAS Number: 375-01-9
MDL Number: MFCD00004674
Molecular Formula: C4H3F7O
Molecular Weight: 200.06
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 25


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Heptafluoro-1-butanol

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 96 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004674
CAS Number: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/05/59/28970559.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR