1,3-Diamino-2-propanol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,3-Diaminopropan-2-ol1,3-Diamino-2-hydroxypropane

H0497-5G H0497-25G
TCH0497-005GEA 40.58 USD
TCH0497-005G TCH0497-025G
1,3-Diamino-2-propanol ≥97.0% (by titrimetric analysis)
1,3-Diamino-2-propanol

CAS Number: 616-29-5
MDL Number: MFCD00008142
Molecular Formula: C3H10N2O
Molecular Weight: 90.13
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Boiling point (°C): 114
Melting point (°C): 42
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Hydroxy-1,3-diaminopropane

Formula: C₃H₁₀N₂O
MW: 90.13 g/mol
Boiling Pt: 114 °C
Melting Pt: 42 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008142
CAS Number: 616-29-5
EINECS: 210-474-2
https://us.vwr.com/stibo/web/std.lang.all/05/07/28970507.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR