1-Hydroxybenzotriazole monohydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: HOBT monohydrate1H-1,2,3-Benzotriazol-1-ol monohydrateHOBT hydrate1-Hydroxybenzotriazole hydrate

H0468-25G
TCH0468-25GEA 74.66 USD
TCH0468-25G
1-Hydroxybenzotriazole monohydrate ≥97.0% (by titrimetric analysis)
1-Hydroxybenzotriazole monohydrate

CAS Number: 80029-43-2
MDL Number: MFCD00005805
Molecular Formula: C6H5N3O
Molecular Weight: 135.13
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 160


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 1,2,3-Benzotriazol-1-ol, HOBt

Formula: C₆H₅N₃O·1H₂O
MW: 153.14 g/mol
Melting Pt: 160 °C
CAS Number: 80029-43-2
EINECS: 219-989-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR