Benzyl-4-hydroxybenzoate ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate
H0209-25G
H0209-100G
TCH0209-025GEA
15.99
USD
TCH0209-025G
TCH0209-100G
Benzyl-4-hydroxybenzoate ≥98.0%
Benzyl 4-hydroxybenzoate
CAS Number: 94-18-8
MDL Number: MFCD00016471
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method:
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Benzyl Ester, Benzylparaben
Formula:
C₁₄H₁₂O₃ Melting Pt: 111 °C |
MDL Number:
MFCD00016471 CAS Number: 94-18-8 EINECS: 202-311-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...