Squalene ≥98.0%

Supplier: TCI America

Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene

H0097-25ML H0097-100ML H0097-500ML
TCH0097-25MLEA 26.9 USD
TCH0097-25ML TCH0097-500ML TCH0097-100ML
Squalene ≥98.0%
Squalene

CAS Number: 111-02-4
MDL Number: MFCD00008912
Molecular Formula: C30H50
Molecular Weight: 410.73
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 252
Melting point (°C): -5
Flash Point (°C): 220
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Triterpenes
Other Product Names / Synonyms: 2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene

Formula: C₃₀H₅₀
Boiling Pt: 252 °C
Melting Pt: –5 °C
Density: 0.86 (20 °C)
Flash Pt: 220 °C
MDL Number: MFCD00008912
CAS Number: 111-02-4
EINECS: 203-826-1
Merck Index: 12,08924
https://us.vwr.com/stibo/web/std.lang.all/01/59/28970159.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR