n-Heptacosane ≥97.0%

Supplier: TCI America

Synonyms: Heptacosane

H0017-10G H0017-1G
TCH0017-010GEA 1142.54 USD
TCH0017-010G TCH0017-1G
n-Heptacosane ≥97.0%
n-Heptacosane

CAS Number: 593-49-7
MDL Number: MFCD00009862
Molecular Formula: C27H56
Molecular Weight: 380.75
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 59
Flash Point (°C): 120

Formula: H₃C(CH₂)₂₅CH₃
MW: 380.74 g/mol
Boiling Pt: 270 °C
Melting Pt: 59 °C
Density: 0.7796 g/cm³ (60 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00009862
CAS Number: 593-49-7
EINECS: 209-792-4
https://us.vwr.com/stibo/web/std.lang.all/00/94/28970094.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR