Glycolic acid ∼70% ∼12 mol/L in water
Supplier: TCI America
|
Danger
|
Synonyms:
2-Hydroxy acid, 2-Hydroxyacetic acid
G0110-500G
G0110-25G
TCG0110-500GEA
37
USD
TCG0110-500G
TCG0110-25G
Glycolic acid ∼70% ∼12 mol/L in water
Glycolic acid
CAS Number: 79-14-1
MDL Number: MFCD00004312
Molecular Formula: C2H4O3
Molecular Weight: 76.05
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 112
Melting point (°C): 10
Specific Gravity (20/20): 1.27
Other Product Names / Synonyms: Hydroxyacetic Acid
Formula:
C₂H₄O₃ MW: 76.05 g/mol Boiling Pt: 266 Melting Pt: 10 °C Density: 1.27 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004312 CAS Number: 79-14-1 EINECS: 201-180-5 UN: 3261 ADR: 8 II Merck Index: 13,04511 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...