L(-)-Glutathione (reduced form) ≥97.0%

Supplier: TCI America

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

G0074-10G G0074-1G
TCG0074-010GEA 94.66 USD
TCG0074-010G TCG0074-001G
L(-)-Glutathione (reduced form) ≥97.0%
L(-)-Glutathione (reduced form)

CAS Number: 70-18-8 MDL Number: MFCD00065939 Molecular Formula: C10H17N3O6S Molecular Weight: 307.32 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 195 Specific rotation [a]20/D: -17 deg (C=2, H2O)


Product Categories: Bioscience, Antioxidants, Oligopeptides Other Product Names / Synonyms: GSH

Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa2e47e0ff59f83fb5ebfc6080ad08598d636c53&vid=fa2e47e0ff59f83fb5ebfc6080ad08598d636c53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR