Glutaraldehyde 24 - 26% in water

Supplier: TCI America
Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

G0067-500ML G0067-25ML
TCG0067-500MLEA 346.84 USD
TCG0067-500ML TCG0067-25ML
Glutaraldehyde 24 - 26% in water
Glutaraldehyde

CAS Number: 111-30-8
MDL Number: MFCD00007025
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.06

Formula: C₅H₈O₂
MW: 100.12
Boiling Pt: 188
Melting Pt: -10
Density: 1.06 (20 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1 II
Merck Index: 13,04485
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c2fbca3c625484220b2ce95b83ea86a8a75bf01&vid=e706d354dac2766116cc4d05eedf7783d53af3a5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR