Furfuryl alcohol ≥98.0%

Supplier: TCI America
Danger

Synonyms: Furfurol

F0076-500ML F0076-25ML
TCF0076-500MLEA 81.99 USD
TCF0076-500ML TCF0076-25ML
Furfuryl alcohol ≥98.0%
Furfuryl alcohol

CAS Number: 98-00-0
MDL Number: MFCD00003252
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Melting point (°C): -29
Flash Point (°C): 78
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 2-Furylcarbinol, 2-(Hydroxymethyl)furan

Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 170 °C
Melting Pt: –29 °C
Density: 1.13 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 13,04326
https://us.vwr.com/stibo/web/std.lang.all/87/08/28968708.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR