EDTA (ethylenediamine tetraacetic acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
EDTA
E0084-500G
E0084-25G
TCE0084-500GEA
96.57
USD
TCE0084-500G
TCE0084-025G
EDTA (ethylenediamine tetraacetic acid) ≥98.0% (by titrimetric analysis)
EDTA (Ethylenediamine tetraacetic acid)
CAS Number: 60-00-4
MDL Number: MFCD00003541
Molecular Formula: C10H16N2O8
Molecular Weight: 292.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 240
Product Categories: Analytical Science, Chelating Agents for Radiopharmaceutical Research, Complexones (Chelating Agents)
Other Product Names / Synonyms: (Ethylenedinitrilo)tetraacetic Acid, EDTA
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol Boiling Pt: 614 °C (1013 hPa) Melting Pt: 240 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...