Ethylenediamine, anhydrous ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane

E0077-500ML E0077-25ML
TCE0077-500MLEA 32.64 USD
TCE0077-500ML TCE0077-25ML
Ethylenediamine, anhydrous ≥98.0%
Ethylenediamine

CAS Number: 107-15-3 MDL Number: MFCD00008204 Molecular Formula: C2H8N2 Molecular Weight: 60.10 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 116 Flash Point (°C): 34 Freezing point (°C): 10 Specific Gravity (20/20): 0.90

Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117
Melting Pt: 8.5
Density: 0.90 (20 °C)
Flash Pt: 34
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8 II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a78a3d2881080e64b9f8dbac699424e45f606ec5&vid=3c52aee6bb207c4f1f21dffe598634973a08861f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR