Ethylene carbonate ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1,3-Dioxa-2-cyclopentanone

E0076-25G E0076-500G
TCE0076-25GEA 21.26 USD
TCE0076-25G TCE0076-500G
Ethylene carbonate ≥99.0%
Ethylene carbonate

CAS Number: 96-49-1 MDL Number: MFCD00005382 Molecular Formula: C3H4O3 Molecular Weight: 88.06 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 244 Melting point (°C): 38 Flash Point (°C): 143 Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Secondary Battery Research Reagents Other Product Names / Synonyms: 1,3-Dioxolan-2-one, 2-Oxo-1,3-dioxolane

Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 243
Melting Pt: 36
Density: 1.47 g/cm³ (20 °C)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00005382
CAS Number: 96-49-1
EINECS: 202-510-0

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR