Ethanesulfonyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 594-44-5
MDL Number: MFCD00007460
Molecular Formula: C2H5ClO2S
Molecular Weight: 128.57
Purity/Analysis Method:
Other Product Names / Synonyms: Ethylsulfonyl Chloride
Formula:
C₂H₅ClO₂S Boiling Pt: 171 °C Melting Pt: –68 °C Density: 1.36 (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007460 CAS Number: 594-44-5 EINECS: 209-842-5 UN: 2927 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...