1,4-Diformylpiperazine
Supplier: TCI America
D2578-25G
D2578-500G
D2578-100G
TCD2578-100GEA
194.82
USD
TCD2578-100G
TCD2578-025G
TCD2578-500G
1,4-Diformylpiperazine
1,4-Diformylpiperazine
CAS Number: 4164-39-0
MDL Number: MFCD00006153
Molecular Formula: C6H10N2O2
Molecular Weight: 142.16
Form: Crystal
Color: White
Melting point (°C): 127
Other Product Names / Synonyms: 1,4-Piperazinedicarboxaldehyde
Formula:
C₆H₁₀N₂O₂ Melting Pt: 127 °C |
MDL Number:
MFCD00006153 CAS Number: 4164-39-0 EINECS: 224-011-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...