2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone2,3-Dichloro-5,6-dicyanoquinone 4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile DDQ

D1070-250G D1070-25G
TCD1070-250GEA 641.3 USD
TCD1070-250G TCD1070-25G
2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥97.0% (by titrimetric analysis)
2,3-Dichloro-5,6-dicyano-p-benzoquinone

CAS Number: 84-58-2
MDL Number: MFCD00001593
Molecular Formula: C8Cl2N2O2
Molecular Weight: 227.00
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 214
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Oxidation, Glycosylation, Materials Chemistry, Molecular Conductors, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: DDQ

Formula: C₈Cl₂N₂O₂
MW: 227.01 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 214 °C
Density: 1.713 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001593
CAS Number: 84-58-2
EINECS: 201-542-2
UN: 3439
ADR: 6.1,III
Merck Index: 13,03088
https://us.vwr.com/stibo/web/std.lang.all/62/95/28976295.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR