1-Dodecanol ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol
D0978-25G
D0978-400G
TCD0978-025GEA
27.16
USD
TCD0978-025G
TCD0978-400G
1-Dodecanol ≥99.0%
1-Dodecanol
CAS Number: 112-53-8
MDL Number: MFCD00004753
Molecular Formula: C12H26O
Molecular Weight: 186.34
Purity/Analysis Method:
Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Dodecyl Alcohol, Lauryl Alcohol
Formula:
H₃C(CH₂)₁₀CH₂OH Boiling Pt: 262 °C Melting Pt: 22…24 °C Density: 0.81976 g/cm³ (20 °C) Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004753 CAS Number: 112-53-8 EINECS: 203-982-0 UN: 3077 ADR: 9,III Merck Index: 13,03439 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...