Dipropylamine ≥99.0%

Supplier: TCI America
Danger

Synonyms: N-Propyl-1-propanamine

D0930-25ML D0930-100ML D0930-500ML
TCD0930-25MLEA 25.8 USD
TCD0930-25ML TCD0930-500ML TCD0930-100ML
Dipropylamine ≥99.0%
Dipropylamine

CAS Number: 142-84-7
MDL Number: MFCD00009362
Molecular Formula: C6H15N
Molecular Weight: 101.19
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Melting point (°C): -63
Flash Point (°C): 7
Specific Gravity (20/20): 0.74

Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 108 °C
Melting Pt: –63 °C
Density: 0.74 (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009362
CAS Number: 142-84-7
EINECS: 205-565-9
UN: 2383
ADR: 3,II
Merck Index: 12,03406
https://us.vwr.com/stibo/web/std.lang.all/60/68/28976068.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR