2,6-Xylenol ≥99.0%

Supplier: TCI America
Danger

Synonyms: 2,6-Dimethylphenol2-Hydroxy-m-xylene2-Hydroxy-3-xylene

D0776-25G D0776-500G
TCD0776-25GEA 19.21 USD
TCD0776-25G TCD0776-500G
2,6-Xylenol ≥99.0%
2,6-Xylenol

CAS Number: 576-26-1
MDL Number: MFCD00002240
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 203
Melting point (°C): 46
Flash Point (°C): 73
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,6-Xylenol

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 203 °C
Melting Pt: 46 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215
https://us.vwr.com/stibo/web/std.lang.all/58/13/28975813.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR