Citral ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3,7-Dimethylocta-2,6-dienal

D0762-25ML
TCD0762-025MLEA 27.96 USD
TCD0762-025ML
Citral ≥98.0%
Citral

CAS Number: 5392-40-5
MDL Number: MFCD00006997
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 104
Specific Gravity (20/20): 0.89


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-2,6-octadienal

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229 °C
Melting Pt: < –10 °C
Density: 0.89 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346
https://us.vwr.com/stibo/web/std.lang.all/57/83/28975783.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR