N,N-Dimethyl-3-nitroaniline ≥99.0%
Supplier: TCI America
|
Warning
|
D0759-25G
TCD0759-025GEA
134
USD
TCD0759-025G
N,N-Dimethyl-3-nitroaniline ≥99.0%
N,N-Dimethyl-3-nitroaniline
CAS Number: 619-31-8
MDL Number: MFCD00007236
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 60
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Boiling Pt: 280 °C Melting Pt: 60 °C Density: 1.313 g/cm³ (17 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007236 CAS Number: 619-31-8 EINECS: 210-590-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...