2-(Dimethylamino)-ethanol ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Dimethylaminoethanol
, N,N-Dimethylethanolamine
, DMAE
CAS Number: 108-01-0
MDL Number: MFCD00002846
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 40
Specific Gravity (20/20): 0.89
Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: N,N-Dimethylethanolamine, 2-Hydroxyethyldimethylamine
Formula:
C₄H₁₁NO MW: 89.14 g/mol Boiling Pt: 134 °C Melting Pt: –59 °C Density: 0.89 (20 °C) Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002846 CAS Number: 108-01-0 EINECS: 203-542-8 UN: 2051 ADR: 8,II Merck Index: 13,02863 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...