2',4'-Dihydroxyacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0561-500G D0561-25G D0561-100G
TCD0561-500GEA 513.44 USD
TCD0561-500G TCD0561-025G TCD0561-100G
2',4'-Dihydroxyacetophenone ≥98.0% (by GC, titration analysis)
2',4'-Dihydroxyacetophenone

CAS Number: 89-84-9 MDL Number: MFCD00002279 Molecular Formula: C8H8O3 Molecular Weight: 152.15 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 145


Other Product Names / Synonyms: 4-Acetylresorcinol, Resacetophenone

Formula: C₈H₈O₃
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 145 °C
Density: 1.18 g/cm³ (141 °C)
MDL Number: MFCD00002279
CAS Number: 89-84-9
EINECS: 201-945-3
Merck Index: 12,08308
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1913c187539d2d39b6d650a3330f6ae90c51085&vid=d1913c187539d2d39b6d650a3330f6ae90c51085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR