Diethylenetriaminepentaacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Kestranal® Aza Pa N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid) Titriplex® V Idranal V DTPA Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid

D0504-25G D0504-500G
TCD0504-500GEA 111.33 USD
TCD0504-500G TCD0504-25G
Diethylenetriaminepentaacetic acid ≥98.0% (by titrimetric analysis)
Diethylenetriaminepentaacetic acid

CAS Number: 67-43-6
MDL Number: MFCD00004289
Molecular Formula: C14H23N3O10
Molecular Weight: 393.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 220


Product Categories: Materials Chemistry, Metal Extractants, Analytical Science, Chelating Agents for Radiopharmaceutical Research, Complexones (Chelating Agents)
Other Product Names / Synonyms: DTPA

Formula: C₁₄H₂₃N₃O₁₀
MW: 393.35 g/mol
Melting Pt: 220 °C
Density: 1.558 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9,III
Merck Index: 12,07266
https://us.vwr.com/stibo/web/std.lang.all/50/89/28975089.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR