α,α'-Dichloro-o-xylene ≥99.0%

Supplier: TCI America

Synonyms: o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride

D0429-500G D0429-25G
TCD0429-500GEA 540 USD
TCD0429-500G TCD0429-25G
α,α'-Dichloro-o-xylene ≥99.0%
α,α'-Dichloro-o-xylene

CAS Number: 612-12-4 MDL Number: MFCD00000903 Molecular Formula: C8H8Cl2 Molecular Weight: 175.05 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 241 Melting point (°C): 55 Flash Point (°C): 108


Other Product Names / Synonyms: o-Xylylene Chloride, o-Xylylene Dichloride

Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 240
Melting Pt: 55 °C
Density: 1.393 g/cm³ (0 °C)
Flash Pt: 108 °C
MDL Number: MFCD00000903
CAS Number: 612-12-4
EINECS: 210-291-8
UN: 2928
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37809d220f3ff24b31c0571a43701b31f237b88d&vid=37809d220f3ff24b31c0571a43701b31f237b88d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR