2,4-Dichlorobenzaldehyde ≥95.0%

Supplier: TCI America

D0330-500G D0330-25G
TCD0330-500GEA 159 USD
TCD0330-500G TCD0330-25G
2,4-Dichlorobenzaldehyde ≥95.0%
2,4-Dichlorobenzaldehyde

CAS Number: 874-42-0 MDL Number: MFCD00003305 Molecular Formula: C7H4Cl2O Molecular Weight: 175.01 Purity/Analysis Method: 95.0% (GC) Form: Crystal Boiling point (°C): 233 Melting point (°C): 72 Flash Point (°C): 135

Formula: C₇H₄Cl₂O
Boiling Pt: 233 °C
Melting Pt: 72 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS Number: 874-42-0
EINECS: 212-861-1
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5b71a599979e6e21bd8ab845d180ffe4cce2f5c&vid=4dda36c6d6e5bad9b48f02a1d0df03c0125c262a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR