Cobalt(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Cobalt dichloride hexahydrate
, Cobalt Chloride, 6-Hydrate
, Cobaltous chloride hexahydrate
, Cobalt Chloride, Hexahydrate
C2388-250G
TCC2388-250GEA
95.94
USD
TCC2388-250G
Cobalt(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Cobalt (II) chloride hexahydrate
CAS Number: 7791-13-1
MDL Number: MFCD00149652
Molecular Formula: Cl2Co
Molecular Weight: 129.83
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 87
Formula:
CoCl₂·6H₂O MW: 237.93 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: 87 °C Density: 1.92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 13,02462 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...