Charcoal activated

Supplier: TCI America

Synonyms: Activated carbonCarbon activatedActivated charcoal

C2194-100G
TCC2194-100GEA 70.5 USD
TCC2194-100G
Charcoal activated
Charcoal activated

[Catalyst for Oxidation using Molecular Oxygen]
CAS Number: 7440-44-0
Molecular Formula: C
Molecular Weight: 12.01
Form: Crystal
Color: Black


Other Product Names / Synonyms: Carbon Activated, Activated Charcoal, Activated Carbon

Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR