Citric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acidHydroxytricarballylic acid

C1949-500G
TCC1949-500GEA 48.76 USD
TCC1949-500G
Citric acid ≥98.0% (by titrimetric analysis)
Citric acid

CAS Number: 77-92-9
Molecular Formula: C6H8O7
Molecular Weight: 192.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 154

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 154 °C
Density: 1.665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://us.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR