Eucalyptol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1,8-Cineole
CAS Number: 470-82-6
MDL Number: MFCD00167977
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 176
Melting point (°C): 2
Flash Point (°C): 48
Specific Gravity (20/20): 0.92
Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: 1,8-Epoxy-p-menthane, Eucalyptol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 176 °C Melting Pt: 2 °C Density: 0.92 (20 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167977 CAS Number: 470-82-6 EINECS: 207-431-5 UN: 3271 ADR: 3,III Merck Index: 12,03940 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...