4-Chloro-2,6-diaminopyrimidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
6-Chloropyrimidine-2,4-diamine
, 2,4-Diamino-6-chloropyrimidine
C0811-25G
TCC0811-025GEA
58.79
USD
TCC0811-025G
4-Chloro-2,6-diaminopyrimidine ≥98.0% (by titrimetric analysis)
4-Chloro-2,6-diaminopyrimidine
CAS Number: 156-83-2
MDL Number: MFCD00006097
Molecular Formula: C4H5ClN4
Molecular Weight: 144.56
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 198
Other Product Names / Synonyms: 2,4-Diamino-6-chloropyrimidine
Formula:
C₄H₅ClN₄ MW: 144.56 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00006097 CAS Number: 156-83-2 EINECS: 205-863-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...