Cyclopentane ≥98.0%

Supplier: TCI America
Danger

Synonyms: Pentamethylene

C0699-25ML C0699-500ML C0699-100ML
TCC0699-100MLEA 49.66 USD
TCC0699-100ML TCC0699-500ML TCC0699-25ML
Cyclopentane ≥98.0%
Cyclopentane

CAS Number: 287-92-3
MDL Number: MFCD00001356
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): -7
Specific Gravity (20/20): 0.75

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 50 °C
Melting Pt: –93.3 °C
Density: 0.75 (20 °C)
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3,II
Merck Index: 13,02769
https://us.vwr.com/stibo/web/std.lang.all/77/49/28937749.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR