Cytidine ≥98.0%

Supplier: TCI America

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

C0522-1G C0522-25G C0522-5G
TCC0522-001GEA 26.87 USD
TCC0522-001G TCC0522-025G TCC0522-5G
Cytidine ≥98.0%
Cytidine

CAS Number: 65-46-3 MDL Number: MFCD00006545 Molecular Formula: C9H13N3O5 Molecular Weight: 243.22 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Specific rotation [a]20/D: 34 deg (C=0.7, H2O)

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006545
CAS Number: 65-46-3
EINECS: 200-610-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a464f934275311983fe32edac4443b279f8db7a6&vid=a464f934275311983fe32edac4443b279f8db7a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR